missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(ethylene glycol), average M.W. 1000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192250051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Average M.W. 1000 | |
25322-68-3 | |
(C2H4O)n | |
MFCD01779601 | |
PEG | |
5 kg | |
[H]OCCO | |
62.07 | |
>260°C | |
Authentic | |
24-29 mPa.s (20°C) |
Poly(ethylene glycol) | |
Waxy Solid | |
35.0°C to 40.0°C | |
H(OCH2CH2)nOH | |
15, 7688 | |
Solubility in water: 750g/L (20°C). Other solubilities: soluble in many organic solvents | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
1000 | |
107 to 118mg KOH/g | |
Plastic bottle | |
0.5% max. (K.F.) |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG | |
[H]OCCO |
Safety and Handling
RTECSNumber : TQ4100000
TSCA : TSCA
RUO – Research Use Only