Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly-D-lysine hydrobromide, M.W. 30,000-70,000

A known nonspecific cell attachment factor

$59.12 - $59.12

Chemical Identifiers

Chemical Name or Material Poly-D-lysine hydrobromide
CAS 27964-99-4
Name Note M.W. 30,000 to 70,000
Formula Weight 30 to 70 kd
MDL Number MFCD00131934
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65578LB0

 
alfa aesar™
J65578LB0
5mg Each for $59.12
Description & Specifications

Specifications

Chemical Name or Material Poly-D-lysine hydrobromide
MDL Number MFCD00131934
CAS 27964-99-4
Name Note M.W. 30,000 to 70,000
Physical Form Solid
Formula Weight 30 to 70 kd
Quantity 5mg
Storage Note 1 2° to 8°C
Recommended Storage 2° to 8°C

Applications
A known nonspecific cell attachment factorPoly-D-lysine hydrobromide is used to culture a wide variety of cell types. It is used as a food additive. Further, it is used in preparing surfaces for cell attachment. In addition to this, it is used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.

Solubility
Soluble in water.

Notes
MW range is 30,000-70,000. Membrane filtered and aseptically lyophilized. Approximately 2µg of poly-D-lysine per square centimeter of plating surface is recommended.Incompatible with strong oxidizing agents.

Safety and Handling

2° to 8°C

2° to 8°C