Please login to your online account to display your discounted pricing

Poly(cyclohexyl methacrylate), approx. Mw 65,000, powder, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  184940100

 View more versions of this product

Catalog No. AC184940100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 10g
Color White
TSCA TSCA
Density 1.1000g/mL
Chemical Name or Material Poly(cyclohexyl methacrylate)
Linear Formula [-CH2C(CH3)(CO2C6H11)-]n
MDL Number MFCD00084408
CAS 25768-50-7
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle

Specifications

Chemical Name or Material Poly(cyclohexyl methacrylate)
CAS 25768-50-7
Linear Formula [-CH2C(CH3)(CO2C6H11)-]n
MDL Number MFCD00084408