Please login to your online account to display your discounted pricing

Poly(cyclohexyl methacrylate), approx. Mw 65,000, powder, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(cyclohexyl methacrylate)
CAS 25768-50-7
Linear Formula [-CH2C(CH3)(CO2C6H11)-]n
MDL Number MFCD00084408
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC184940100

  SDS
thermo fisher scientific
184940100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 10g
Color White
TSCA TSCA
Density 1.1000g/mL
Chemical Name or Material Poly(cyclohexyl methacrylate)
Linear Formula [-CH2C(CH3)(CO2C6H11)-]n
MDL Number MFCD00084408
CAS 25768-50-7
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle