Please login to your online account to display your discounted pricing

Poly(caprolactone) triol, average M.W. 900, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  190755000

 View more versions of this product

Catalog No. AC190755000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Paste
Quantity 500g
Color White
Density 1.0800g/mL
Chemical Name or Material Poly(caprolactone) triol
Name Note Average M.W. 900
MDL Number MFCD00084405
CAS 54735-63-6
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle

Specifications

Chemical Name or Material Poly(caprolactone) triol
Name Note Average M.W. 900
CAS 54735-63-6
MDL Number MFCD00084405