Please login to your online account to display your discounted pricing

Poly(caprolactone) triol, average M.W. 900, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(caprolactone) triol
Name Note Average M.W. 900
CAS 54735-63-6
MDL Number MFCD00084405
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190755000

  SDS
thermo fisher scientific
190755000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Paste
Quantity 500g
Color White
Density 1.0800g/mL
Loss on Drying 5% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note Average M.W. 900
MDL Number MFCD00084405
CAS 54735-63-6
Chemical Name or Material Poly(caprolactone) triol