missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(caprolactone) triol, average M.W. 300, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC190730250

 View more versions of this product

Catalog No. AC190730250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Viscous Liquid
Clear colorless
1.4750 to 1.4780 (20°C, 589nm)
Glass bottle
MFCD00084405
Poly(caprolactone) triol
25g
1.0700g/mL
Authentic
2% max.
39280-09-6

Chemical Identifiers

39280-09-6
MFCD00084405
Poly(caprolactone) triol