Please login to your online account to display your discounted pricing

Poly(caprolactone) Diol, Average M.W. 2000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(caprolactone) diol
CAS 36890-68-3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183212500

  SDS
fisher scientific acros
183212500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC183215000

  SDS
fisher scientific acros
183215000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC183210025

  SDS
thermo fisher scientific
AC183210025
Plastic Bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Waxy Solid
Color White
Quantity 250g
Density 1.0700g/mL
Chemical Name or Material Poly(caprolactone) diol
CAS 36890-68-3
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass bottle