missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 184990050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly(acrylic acid) | |
Liquid | |
100.0°C | |
[-CH2CH(CO2H)-]n | |
1.2300g/mL | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane | |
NNMHYFLPFNGQFZ-UHFFFAOYSA-M | |
Poly(acrylic acid) | |
6581 | |
60.0 to 65.0% (solids) | |
Glass bottle |
63 wt. % Aqueous Solution | |
9003-01-4,79-10-7,7732-18-5 | |
(C3H4O2)n | |
MFCD00084394 | |
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
5 g | |
OC(=O)C(-*)C-* | |
72.06 | |
CHEBI:18308 | |
Authentic | |
1.23 |
Chemical Identifiers
9003-01-4 | |
72.06 | |
NNMHYFLPFNGQFZ-UHFFFAOYSA-M | |
6581 | |
OC(=O)C(-*)C-* |
(C3H4O2)n | |
MFCD00084394 | |
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
CHEBI:18308 |
Safety and Handling
RTECSNumber : AT4680000
TSCA : TSCA
RUO – Research Use Only