Please login to your online account to display your discounted pricing

Poly(acrylic acid), 50 wt.% solution in water; ∽ M.W. 5,000, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  185000050

 View more versions of this product

Catalog No. AC185000050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Quantity 5g
Color Clear to hazy
TSCA TSCA
Density 1.1800g/mL
Assay 48 to 52% (solids)
RTECS Number AT4680000
Chemical Name or Material Poly(acrylic acid)
Name Note 50 wt. % Solution in Water
Linear Formula [-CH2CH(CO2H)-]n
MDL Number MFCD00084394
CAS 9003-01-4
Health Hazard 1
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 7732-18-5

Specifications

Chemical Name or Material Poly(acrylic acid)
Name Note 50 wt. % Solution in Water
CAS 9003-01-4
CAS 7732-18-5
Assay 48 to 52% (solids)
Linear Formula [-CH2CH(CO2H)-]n
MDL Number MFCD00084394
Accessories

SDS & Safety Documents