Learn More
Poly(acrylic acid), 25 wt% soln. in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04466936
Description
Polyacrylic acid is used in disposable diapers and in ion exchange resins. It is also used to study solute diffusion in polyvinyl alcohol/polyacrylic acid copolymer hydrogel. It is also employed as a thickening, suspending, emulsifying and dispersing agent in pharmaceuticals, cosmetics, adhesives and paints. Further, it is used for the preparation of poly(N-isopropylacrylamide)-block-polyacrylic acid copolymer which responds to both temperature and pH stimuli. In addition to this, it is used in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly (acrylic acid) | |
9003-01-4 | |
MFCD00084394 | |
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
500 g | |
OC(=O)C(-*)C-* | |
72.06 | |
CHEBI:18308 |
Liquid | |
(C3H4O2)n | |
1.09 g/mL | |
Soluble in water. | |
NNMHYFLPFNGQFZ-UHFFFAOYSA-M | |
Poly(acrylic acid) | |
6581 | |
25 wt% soln. in water |
Chemical Identifiers
9003-01-4 | |
72.06 | |
NNMHYFLPFNGQFZ-UHFFFAOYSA-M | |
6581 | |
Poly(acrylic acid) |
(C3H4O2)n | |
MFCD00084394 | |
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
CHEBI:18308 | |
OC(=O)C(-*)C-* |
Safety and Handling
RTECSNumber : AT4680000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only