missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 178045000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White to Yellow | |
Granules | |
(C3H5NO)n | |
MFCD00084392 | |
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer | |
500 g | |
NC(=O)C(-*)C-* | |
72.06 | |
CHEBI:18308 | |
Authentic | |
Glass bottle |
Poly(acrylamide) | |
9003-05-8 | |
[-CH2CH(CONH2)-]n | |
1.3000g/mL | |
Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enoic acid | |
6581 | |
5 to 6.000.000 (Average ) | |
13% max. | |
1.3 |
Chemical Identifiers
9003-05-8 | |
MFCD00084392 | |
NC(=O)C(-*)C-* |
(C3H5NO)n | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N |
Safety and Handling
RTECSNumber : AS3700000
TSCA : TSCA
RUO – Research Use Only