Please login to your online account to display your discounted pricing

Poly(acrylamide), carboxyl modified, pure, low carboxyl content, ∽ MW 200,000, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  184232500

 View more versions of this product

Catalog No. AC184232500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight ≈200.000
Physical Form Powder or Granules
Quantity 250g
Color White
Purity Grade Pure
Loss on Drying 10% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Additional Information linear polymer
Name Note Carboxyl Modified, Low Carboxyl Conten
CAS 62649-23-4
Chemical Name or Material Poly(acrylamide), carboxyl modified
MDL Number MFCD00084393

Specifications

Chemical Name or Material Poly(acrylamide), carboxyl modified
Name Note Carboxyl Modified, Low Carboxyl Conten
CAS 62649-23-4
Purity Grade Pure
Formula Weight ≈200.000
MDL Number MFCD00084393