Please login to your online account to display your discounted pricing

Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  184242500

 View more versions of this product

Catalog No. AC184242500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Granules
Quantity 250g
Color Off-white
Purity Grade Pure
Chemical Name or Material Poly(acrylamide), carboxyl modified
MDL Number MFCD00084393
CAS 62649-23-4
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle

Specifications

Chemical Name or Material Poly(acrylamide), carboxyl modified
CAS 62649-23-4
Purity Grade Pure
MDL Number MFCD00084393