Please login to your online account to display your discounted pricing

Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, ACROS Organics™

$94.87 - $94.87

Chemical Identifiers

Chemical Name or Material Poly(acrylamide), carboxyl modified
CAS 62649-23-4
Purity Grade Pure
MDL Number MFCD00084393
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC184240050

  SDS
thermo fisher scientific
184240050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC184242500

  SDS
thermo fisher scientific
184242500
Glass bottle 250g Each for $94.87
Description & Specifications

Specifications

Physical Form Granules
Quantity 5g
Color Off-white
Purity Grade Pure
Packaging Glass bottle
Loss on Drying 10% max.
Infrared Spectrum Authentic
Chemical Name or Material Poly(acrylamide), carboxyl modified
CAS 62649-23-4
MDL Number MFCD00084393