Please login to your online account to display your discounted pricing

Poly(acrylamide), carboxyl modified, Acros Organics

$120.04 - $120.04

Chemical Identifiers

Chemical Name or Material Poly(acrylamide), carboxyl modified
CAS 62649-23-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC448530050

  SDS
thermo fisher scientific
AC448530050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC448532500

  SDS
thermo fisher scientific
AC448532500
Glass bottle 250g Each for $120.04
Description & Specifications

Specifications

Quantity 5g
Packaging Glass bottle
Chemical Name or Material Poly(acrylamide), carboxyl modified
CAS 62649-23-4