Please login to your online account to display your discounted pricing

Poly(4-vinylpyridine), 2% cross-linked with DVB, ca. 60 mesh, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  210542500

 View more versions of this product

Catalog No. AC210542500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 250g
Color White to cream
Fieser 16,282
TSCA TSCA
MDL Number MFCD00190588
Additional Information base capacity: ca. 8.0 meq/g (dry)
Chemical Name or Material Poly(4-vinylpyridine)
CAS 9017-40-7
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% max.

Specifications

Chemical Name or Material Poly(4-vinylpyridine)
CAS 9017-40-7
MDL Number MFCD00190588