Please login to your online account to display your discounted pricing

Poly(4-vinylpyridine), 2% cross-linked with DVB, ca. 60 mesh, ACROS Organics™

$463.19 - $463.19

Chemical Identifiers

Chemical Name or Material Poly(4-vinylpyridine)
CAS 9017-40-7
MDL Number MFCD00190588
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC210542500

  SDS
thermo fisher scientific
210542500
Plastic bottle 250g Each for $463.19
Description & Specifications

Specifications

Physical Form Powder
Quantity 250g
Color White to cream
Fieser 16,282
TSCA TSCA
Additional Information base capacity: ca. 8.0 meq/g (dry)
MDL Number MFCD00190588
Infrared Spectrum Authentic
Water 10% max.
Packaging Plastic bottle
Chemical Name or Material Poly(4-vinylpyridine)
CAS 9017-40-7