Please login to your online account to display your discounted pricing

Poly(4-methyl-1-pentene), low molecular weight, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(4-methyl-1-pentene)
CAS 25068-26-2
Linear Formula {-CH2CHCH2CH(CH3)2-}n
MDL Number MFCD00084382
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC184285000

  SDS
thermo fisher scientific
184285000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Quantity 500g
Color Clear
TSCA TSCA
Density 0.83g/mL
Chemical Name or Material Poly(4-methyl-1-pentene)
Linear Formula {-CH2CHCH2CH(CH3)2-}n
MDL Number MFCD00084382
CAS 25068-26-2
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle