Please login to your online account to display your discounted pricing

Poly(4-chlorostyrene), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC178350010

 View more versions of this product

Catalog No. AC178350010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 51000
Quantity 1g
Color White
Density 1.5500g/mL
Loss on Drying 2% max.
Packaging Glass bottle
Chemical Name or Material Poly(4-chlorostyrene)
Linear Formula [-CH2CH(C6H4Cl)-]n
MDL Number MFCD00084381
CAS 24991-47-7
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Poly(4-chlorostyrene)
CAS 24991-47-7
Linear Formula [-CH2CH(C6H4Cl)-]n
Formula Weight 51000
MDL Number MFCD00084381