Please login to your online account to display your discounted pricing

Poly(4-chlorostyrene), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(4-chlorostyrene)
CAS 24991-47-7
Linear Formula [-CH2CH(C6H4Cl)-]n
Formula Weight 51000
MDL Number MFCD00084381
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC178350010

  SDS
thermo fisher scientific
AC178350010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC178350050

  SDS
thermo fisher scientific
178350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 51000
Physical Form Powder
Quantity 1g
Color White
Density 1.5500g/mL
Loss on Drying 2% max.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 24991-47-7
Chemical Name or Material Poly(4-chlorostyrene)
Linear Formula [-CH2CH(C6H4Cl)-]n
MDL Number MFCD00084381