Alfa Aesar™ Poly(3-octylthiophene-2,5-diyl), regiorandom, low metals
Conducting polymers
Manufacturer:  Alfa Aesar™ 4477803
 View more versions of this product
Catalog No. AA4477803
Specifications
Specifications
Poly (3-octylthiophene-2,5-diyl)
104934-51-2
MFCD00241412
Soluble in THF, CHCl3
CCCCCCCCC1=C(SC(=C1)C)C.N
241.437
Powder
Regiorandom, low metals
C14H27NS
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
RXIQSINDOTYIGV-UHFFFAOYSA-N
azane;2,5-dimethyl-3-octylthiophene
121233643
1g
Chemical Identifiers
104934-51-2
241.437
RXIQSINDOTYIGV-UHFFFAOYSA-N
121233643
CCCCCCCCC1=C(SC(=C1)C)C.N
C14H27NS
MFCD00241412
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
azane;2,5-dimethyl-3-octylthiophene
SDS Documents