Learn More
Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04474577
Description
Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) and devices. It is also used as organic light emitting diode (OLED) materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly (3-hexylthiophene-2,5-diyl) | |
Powder | |
(C10H14S)n | |
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity | |
0.1 g | |
CCCCCCC1=C(-*)SC(-*)=C1 | |
NaN |
Regioregular, low metals | |
104934-50-1 | |
MFCD00217686 | |
Soluble in Chloroform and tetrahydrofuran | |
DUFPJSOXRHVDOV-UHFFFAOYSA-N | |
3-hexyl-2,5-dimethylthiophene | |
22950249 |
Chemical Identifiers
104934-50-1 | |
NaN | |
DUFPJSOXRHVDOV-UHFFFAOYSA-N | |
22950249 | |
CCCCCCC1=C(-*)SC(-*)=C1 |
(C10H14S)n | |
MFCD00217686 | |
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity | |
3-hexyl-2,5-dimethylthiophene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only