Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(3-butylthiophene-2,5-diyl), regioregular

$284.77 - $284.77

Chemical Identifiers

Chemical Name or Material Poly (3-butylthiophene-2,5-diyl)
CAS 98837-51-5
Name Note Regioregular
Molecular Formula (C8H12S)n
MDL Number MFCD00677712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4478701

 
alfa aesar™
4478701
0.25g Each for $284.77
Description & Specifications

Specifications

Chemical Name or Material Poly (3-butylthiophene-2,5-diyl)
Name Note Regioregular
Physical Form Powder
Molecular Formula (C8H12S)n
MDL Number MFCD00677712
CAS 98837-51-5
Color Yellow to green
Quantity 0.25g