Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(3-butylthiophene-2,5-diyl), regiorandom

$257.05 - $257.05

Chemical Identifiers

Chemical Name or Material Poly (3-butylthiophene-2,5-diyl)
CAS 98837-51-5
Name Note Regiorandom
Molecular Formula (C8H12S)n
MDL Number MFCD00677712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4478801

 
alfa aesar™
4478801
0.25g Each for $257.05
Description & Specifications

Specifications

Chemical Name or Material Poly (3-butylthiophene-2,5-diyl)
Name Note Regiorandom
Physical Form Powder
Molecular Formula (C8H12S)n
MDL Number MFCD00677712
CAS 98837-51-5
Color Brown to black
Quantity 0.25g