missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(2-methylaniline), emeraldine salt from p-toluenesulfonic acid

$49.72 - $49.72

Chemical Identifiers

Molecular Formula −[C6H3(CH3)NH-]n·(C6H4CH3SO3)m
MDL Number MFCD02688597
Chemical Name or Material Poly (2-methylaniline), Emeraldine salt from p-toluenesulfonic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4380606 SDS Alfa Aesar™
4380606
5g Each for $49.72
AA4380614 SDS Alfa Aesar™
4380614
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4380622 SDS Alfa Aesar™
4380622
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

−[C6H3(CH3)NH-]n·(C6H4CH3SO3)m
MFCD02688597
Poly (2-methylaniline), Emeraldine salt from p-toluenesulfonic acid

Specifications

Powder
MFCD02688597
Poly (2-methylaniline), Emeraldine salt from p-toluenesulfonic acid
−[C6H3(CH3)NH-]n·(C6H4CH3SO3)m
5g