Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(2-methylaniline), emeraldine base

$241.74 - $1,326.38

Chemical Identifiers

Chemical Name or Material Poly (2-methylaniline)
Name Note Emeraldine base
Molecular Formula −[C6H3(CH3)NH-]n
MDL Number MFCD02688596
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4380522

 
alfa aesar™
43805-22
100g Each for $241.74

AA4380536

 
alfa aesar™
43805-36
500g Each for $1,326.38
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Poly (2-methylaniline)
Name Note Emeraldine base
Physical Form Powder
Molecular Formula −[C6H3(CH3)NH-]n
MDL Number MFCD02688596