Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(2-methoxyaniline), emeraldine salt from hydrochloric acid

$170.66 - $170.66

Chemical Identifiers

Chemical Name or Material Poly (2-methoxyaniline)
Name Note Emeraldine salt from hydrochloric acid
Molecular Formula −[C6H3(CH3O)NH]-n·HCl
MDL Number MFCD02688595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4384022

 
alfa aesar™
43840-22
100g Each for $170.66
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Poly (2-methoxyaniline)
Name Note Emeraldine salt from hydrochloric acid
Physical Form Powder
Molecular Formula −[C6H3(CH3O)NH]-n·HCl
MDL Number MFCD02688595
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.