Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(2-methoxyaniline), emeraldine base

$142.24 - $142.24

Chemical Identifiers

Chemical Name or Material Poly (2-methoxyaniline)
Name Note Emeraldine base
Molecular Formula −[C6H3(CH3O)NH]-n
MDL Number MFCD02688594
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4384106

  SDS
alfa aesar™
4384106
5g Each for $142.24
Description & Specifications

Specifications

Chemical Name or Material Poly (2-methoxyaniline)
Name Note Emeraldine base
Physical Form Powder
Molecular Formula −[C6H3(CH3O)NH]-n
MDL Number MFCD02688594
Quantity 5g