Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(2-ethylaniline), emeraldine base

Preparation of conductive polymers

$26.58 - $26.58

Chemical Identifiers

Chemical Name or Material Poly (2-ethylaniline)
Name Note Emeraldine base
Molecular Formula −[C6H3(C2H5)NH-]n
MDL Number MFCD02688592
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4382506

 
alfa aesar™
43825-06
5g Each for $26.58
Description & Specifications

Specifications

Quantity 5g
Color Dark blue to black
Quantity 5g
Chemical Name or Material Poly (2-ethylaniline)
Name Note Emeraldine base
Physical Form Powder
Molecular Formula −[C6H3(C2H5)NH-]n
MDL Number MFCD02688592
Solubility Information Soluble in organic solvents