Learn More
Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, ACROS Organics™
Chemical Identifiers
CAS | 27119-07-9 |
---|---|
Molecular Formula | C7H13NO4S |
Molecular Weight (g/mol) | 207.244 |
MDL Number | MFCD00084369 |
InChI Key | XHZPRMZZQOIPDS-UHFFFAOYSA-N |
Synonym | 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, 2-acrylamide-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methylpropanesulphonic acid, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5 |
PubChem CID | 65360 |
IUPAC Name | 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid |
SMILES | CC(C)(CS(=O)(=O)O)NC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC184960250
|
Thermo Fisher Scientific
184960250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC184961000
|
Thermo Fisher Scientific
184961000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC184965000
|
Thermo Fisher Scientific
AC184965000 |
500g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
27119-07-9 | |
207.244 | |
XHZPRMZZQOIPDS-UHFFFAOYSA-N | |
65360 | |
CC(C)(CS(=O)(=O)O)NC(=O)C=C |
C7H13NO4S | |
MFCD00084369 | |
1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, 2-acrylamide-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methylpropanesulphonic acid, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5 | |
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid |
Specifications
27119-07-9,7732-18-5 | |
{-CH2CH[CONHC(CH3)2CH2SO3H]-}n | |
1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, 2-acrylamide-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methylpropanesulphonic acid, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5 | |
CC(C)(CS(=O)(=O)O)NC(=O)C=C | |
207.244 | |
Authentic | |
8.0 to 12.0% (solids) | |
Undesignated | |
Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution |
C7H13NO4S | |
MFCD00084369 | |
XHZPRMZZQOIPDS-UHFFFAOYSA-N | |
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid | |
65360 | |
Viscous Liquid | |
Glass bottle | |
25g |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning