Please login to your online account to display your discounted pricing

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  184961000

 View more versions of this product

Catalog No. AC184961000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Quantity 100g
Color Clear
Assay 8.0 to 12.0% (solids)
Name Note 10 wt% aqueous solution
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS 7732-18-5
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid)
MDL Number MFCD00084369
CAS 27119-07-9
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid)
Name Note 10 wt% aqueous solution
CAS 27119-07-9
CAS 7732-18-5
Assay 8.0 to 12.0% (solids)
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
MDL Number MFCD00084369
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents