Argos Technologies™ PolarSafe™ ICE Buckets and Pans
Ice buckets and pans have excellent insulating properties to help keep temperature sensitive samples safely chilled.
$90.67 - $110.17
Specifications
Material Polystyrene
For Use With (Application) Ice, dry ice, slurry ice
View More Specs
Products 3
Catalog Number Mfr. No. Type Capacity (Metric) Color Price Quantity  
Catalog Number Mfr. No. Type Capacity (Metric) Color Price Quantity  
10-987-060
View Documents
Argos Technologies™
0439694
Ice Bucket 2.5 L Black
Each for $90.67
10-987-061
View Documents
Argos Technologies™
0439695
Ice Bucket 5 L Blue
Each for $110.17
10-987-063
View Documents
Argos Technologies™
0439697
Ice Pan 4 L Blue
Each for $90.67
Description
Description
  • High density expanded polystyrene construction provides extra rigidity and strength, and to prevent condensation on benchtops
  • Buckets come with recessed handle and snug fitting lids with easy to grab knob to minimize evaporation loss and to maximize cooling
  • Pans have gripping ridges for easy transportation and pour spout to empty cooling material when no longer needed
Specifications
Specifications
Polystyrene
Ice, dry ice, slurry ice
SDS