missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Argos Technologies™ PolarSafe™ Cryogenic Storage Vials with Internal Cap
Tubes are available with round or self-standing bottoms
$70.12 - $90.82
Specifications
Temperature Range (Metric) | -196°C to +121°C |
---|---|
Thread Style | Internal |
RNase Free | Yes |
Dnase Free | Yes |
Barcode | Serial/consecutive Barcode128 printed on each tube |
Catalog Number | Mfr. No. | Interior Bottom Shape | Height (Metric) | Volume (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Interior Bottom Shape | Height (Metric) | Volume (Metric) | Price | Quantity | |||
03-391-528
|
Argos Technologies™
0439564 |
Self-standing | 43 mm | 1 mL |
Case of 50 Each for $70.12
|
|
03-391-529
|
Argos Technologies™
0439565 |
Self-standing | 48 mm | 2 mL |
Case of 50 Each for $73.29
|
|
03-391-530
|
Argos Technologies™
0439566 |
Round | 47 mm | 2 mL |
Case of 50 Each for $70.12
|
|
03-391-531
|
Argos Technologies™
0439567 |
Self-standing | 76 mm | 4 mL |
Case of 50 Each for $77.26
|
|
03-391-532
|
Argos Technologies™
0439568 |
Round | 75 mm | 4 mL |
Pack of 50 Each for $90.82
|
|
03-391-533
|
Argos Technologies™
0439569 |
Self-standing | 90 mm | 5 mL |
Case of 50 Each for $80.47
|
|
03-391-534
|
Argos Technologies™
0439570 |
Round | 90 mm | 5 mL |
Case of 50 Each for $90.82
|
|
Description
Space-saving design with our smallest thermal cycler footprint
- Flexible modular design saves space on deck
- The separate control panel module provides flexibility when integrating on the robotic deck
- Choice of 10cm,1 meter and 3 meter cabling options
Automated lid and cycler features
- 384 well block
- The automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
- The heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation
Do not use vials for storage in the liquid phaes of liquid nitrogen: this may cause entrapment of liquified nitrogen inside the vial and lead to pressure build-up,resulting in possible explosion or biohazard release.
Specifications
-196°C to +121°C | |
Yes | |
Serial/consecutive Barcode128 printed on each tube | |
Autoclavable |
Internal | |
Yes | |
Polyethylene, Thermoplastic Elastomer | |
FDA, GMP and ISO quality standards (ISO13485:2004, ISO14644 & 14698) |