United Scientific Supplies Polarizing Films
An effective yet economical tool for demonstrating polarization of light.
$12.48 - $41.08
Specifications
Product Type Polarizing Film
For Use With (Application) Ideal for introducing and demonstrating the polarization of light
View More Specs
Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
S07373 View Documents United Scientific Supplies
PLF030
3 in. 7.62 cm 3 in. 7.62cm Each for $12.48
S07374 View Documents United Scientific Supplies
PLF060
6 in. 15.2cm 6 in. 15.2cm Each for $41.08
Description
Description
  • Available mounted or unmounted
  • Three sizes: 2 × 2 in. (5 × 5cm), 3 × 3 in. (7.6 × 7.6cm), 6 × 6 in. (15.2 × 15.2cm))
Order Info
2 × 2 in. films are available in packs of 10 and 3 × 3 in. and 6 × 6 in. films are sold individually.
Specifications
Specifications
Polarizing Film
Ideal for introducing and demonstrating the polarization of light