Polarizing Film
Demonstrate that light is a transverse wave.
Manufacturer:  Fisher Scientific IFPF2
 View more versions of this product
Catalog No. S83161
Description
Description
  • General-purpose linear polarizing film is useful in visible and near-infrared light
  • Extinction ratio (the ratio of minimum to maximum intensity) is greater than 0.01%
  • Use in optical experiments such as verification of the Law of Malus and light polarization
  • Easy to cut, punch or drill
  • Resistant to heat, impact and abrasion
  • Thickness: 0.02 in. (0.5mm)
Specifications
Specifications
Use in optical experiments such as verification of the law of malus and light polarization
100/Pk.
0.5mm Thick Polarizing Film
50mm
50mm
Documents