Please login to your online account to display your discounted pricing

Pokemon (LRF) Armenian Hamster anti-Human, eFluor 660, Clone: 13E9, eBioscience™

Armenian Hamster Monoclonal Antibody

$191.40 - $442.25

Specifications

Antigen Pokemon (LRF)
Clone 1.30E+10
Host Species Armenian Hamster
Gene Alias Leukemia/lymphoma-related factor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124338

 
life technologies
LS50330982
100μg Each for $442.25

501124337

 
life technologies
LS50330980
25μg Each for $191.40
Description & Specifications

Specifications

Antigen Pokemon (LRF)
Clone 1.30E+10
Host Species Armenian Hamster
Gene Alias Leukemia/lymphoma-related factor
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This monoclonal antibody reacts with human Leukemia/lymphoma-related factor (LRF). This 65kDa protein is also known as POKEMON, FBI-1, and Zbtb7a. LRF is a member of the POK family of transcriptional repressors, and is expressed in hematopoietic stem cells, T and B lymphocytes, macrophages, and erythroid cells. This protein is also highly expressed in human non-Hodgkin’s lymphoma tissues and diffuse large B-cell lymphomas, demonstrating its prominent role in oncogenic transformation. Studies have demonstrated that LRF is also involved in B cell lineage commitment and development by opposing Notch signaling. Moreover, LRF is critical for erythroid terminal differentiation in the bone marrow by inhibiting Bim-mediated apoptosis. Finally, LRF interacts with Bcl-6 in germinal center B cells.