Please login to your online account to display your discounted pricing

Podoplanin Golden Syrian Hamster anti-Mouse, PE-Cy7, Clone: eBio8.1.1 (8.1.1), eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen Podoplanin
Clone eBio8.1.1 (8.1.1)
Host Species Golden Syrian Hamster
Gene Alias Pdpn
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123378

 
life technologies
LS25538180
25μg Each for $172.55

501123379

 
life technologies
LS25538182
100μg Each for $433.55
Description & Specifications

Specifications

Antigen Podoplanin
Clone eBio8.1.1 (8.1.1)
Host Species Golden Syrian Hamster
Gene Alias Pdpn
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 8.1.1 monoclonal antibody reacts with mouse podoplanin (T1a, gp38, aggrus), a 43 kDa transmembrane glycoprotein, named for its expression in kidney glomerular epithelial cells (podocytes). In addition, Podoplanin is expressed in epithelial and mesothelial cells such as intestinal epithelium, alveolar type I cells, podocytes, and mesothelium of the visceral peritoneum. It was also shown to be a potent marker for lymphatic endothelium. Podoplanin is also expressed by subcapsular epithelial cells of the murine thymus. Mice deficient in Podoplanin die at birth because of a respiratory defect and congenital lymphedema due to a failure in lymphatic pattern formation.