missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Podocalyxin Mouse anti-Human, PE, Clone: B34D1.3, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $315.48

Specifications

Antigen Podocalyxin
Clone B34D1.3
Host Species Murine
Gene Alias Gp200
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245532 Life Technologies
LS12887341
25 Tests Each for $157.08
50245533 Life Technologies
LS12887342
100 Tests Each for $315.48
Specifications

Description

The monoclonal antibody B34D1.3 recognizes human podocalyxin, a protein expressed on undifferentiated human embryonic stem cells (ES), embryonic carcinoma cells (EC), and embryonic germ cells (EG). Podocalxyin is a heavily glycosylated cell adhesion protein with homology to CD34 and endoglycan. It was first identified on kidney podocytes, and then found to be upregulated in a variety of human cancers such as breast and prostate cancer. Like other stem cell-specific markers, the epitope recognized by the B34D1.3 antibody is lost upon cell differentiation.

The B34D1.3 antibody recognizes the ectodomain of podocalyxin, which is a different epitope from the Tra-1-60 and Tra-1-81 monoclonal antibodies. While Tra1-60 has been found to react with a sialidase-sensitive epitope, the epitope recognized by Tra1-81 has not been fully defined.
Description & Specifications

Specifications

Podocalyxin
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B34D1.3
Gp200
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test