Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.
$1,328.06 - $6,362.06
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Sterility Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Packaging Sterility Volume (Metric) Price Quantity  
14-387-583 Thermo Scientific™
MX5696100
20 racks of 96 tips/case Nonsterile 100nL Case for $1,328.06
14-387-587 Thermo Scientific™
MX5697250
20 racks of 96 tips/case Sterile 250nL Case for $1,601.89
14-387-589 Thermo Scientific™
MX5756100
20 Racks of 384 Tips/Case Nonsterile 100nL Case for $5,502.09
14-387-590 Thermo Scientific™
MX5756250
20 Racks of 384 Tips/Case Nonsterile 250nL Case for $5,910.23
14-387-593 Thermo Scientific™
MX5757250
20 Racks of 384 Tips/Case Sterile 250nL Case for $6,362.06
14-387-594 Thermo Scientific™
MX575750
20 Racks of 384 Tips/Case Sterile 50nL Case of 50 Each for $5,928.00
14-387-584 Thermo Scientific
MX5696250
20 racks of 96 tips/case Nonsterile 250nL Case of 250 Each for $1,328.06
14-387-585 Thermo Scientific
MX569650
20 racks of 96 tips/case Nonsterile 50nL Case of 50 Each for $1,328.06
14-387-586 Thermo Scientific
MX5697100
20 racks of 96 tips/case Sterile 100nL Case of 100 Each for $1,601.89
14-387-588 Thermo Scientific
MX569750
20 racks of 96 tips/case Sterile 50nL Case of 50 Each for $1,601.89
14-387-591 Thermo Scientific™
MX575650
20 Racks of 384 Tips/Case Nonsterile 50nL Case of 50 Each for $5,502.09
14-387-592 Thermo Scientific
MX5757100
20 Racks of 384 Tips/Case Sterile 100nL Case of 100 Each for $5,928.00
Specifications
Description
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.
Features:
  • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
  • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
  • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
  • Control consistent accuracy and precision during the molding process
Validation and Compatibility:
  • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
  • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
  • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
  • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
Ordering Information:
  • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
  • Sterile format available for sensitive liquid handling procedures
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.