Please login to your online account to display your discounted pricing

PNU 74654, Tocris™

β-catenin binder; inhibits Wnt signaling

$179.63 - $750.85

Chemical Identifiers

Chemical Name or Material PNU 74654
Assay Percent Range >99%
Molecular Formula C19H16N2O3
Formula Weight 320.34
Synonym 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

353450

 
r & d systems
RDS353450
50mg Each for $750.85

353410

 
r & d systems
RDS353410
10mg Each for $179.63
Description & Specifications

Specifications

Formula Weight 320.34
Quantity 50mg
CAS 113906-27-7
Synonym 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
Chemical Name or Material PNU 74654
Assay Percent Range >99%
Molecular Formula C19H16N2O3
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C