Please login to your online account to display your discounted pricing

PNU 282987, Tocris

Selective α7 nAChR agonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material PNU 282987
Assay Percent Range >98%
Molecular Formula C14H17ClN2O
Formula Weight 264.75
Synonym N-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-4-chlorobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

230350

 
r & d systems
RDS230350
50mg Each for $794.96

230310

 
r & d systems
RDS230310
10mg Each for $188.60
Description & Specifications

Specifications

Chemical Name or Material PNU 282987
CAS 123464-89-1
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C14H17ClN2O
Formula Weight 264.75
Synonym N-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-4-chlorobenzamide
Quantity 50mg
Safety and Handling

Store at Room Temperature