Please login to your online account to display your discounted pricing

PNU 177864 hydrochloride, Tocris

Highly selective D3 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material PNU 177864 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H21F3N2O3S·HCl
Formula Weight 438.89
Synonym N-[4-[2-(Propylamino)ethyl)phenyl]-4-(trifluoromethoxy)-benzenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

335550

 
r & d systems
RDS335550
50mg Each for $844.73

335510

 
r & d systems
RDS335510
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 438.89
Quantity 50mg
Chemical Name or Material PNU 177864 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H21F3N2O3S·HCl
CAS 250266-51-4
Synonym N-[4-[2-(Propylamino)ethyl)phenyl]-4-(trifluoromethoxy)-benzenesulfonamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature