Please login to your online account to display your discounted pricing

PMPA (NMDA antagonist), Tocris

Competitive NMDA antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material PMPA (NMDA antagonist)
Assay Percent Range >98%
Molecular Formula C6H13N2O5P
Formula Weight 224.15
Synonym 4-(Phosphonomethyl)-2-piperazinecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

227310

 
r & d systems
227310
10mg Each for $239.63

227350

 
r & d systems
227350
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 224.15
Quantity 10mg
Chemical Name or Material PMPA (NMDA antagonist)
CAS 113919-36-1
Synonym 4-(Phosphonomethyl)-2-piperazinecarboxylic acid
Assay Percent Range >98%
Molecular Formula C6H13N2O5P
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C