Please login to your online account to display your discounted pricing

PluriSln 1, Tocris™

Inhibitor of SCD1; selectively eliminates undifferentiated hPSCs from culture

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material PluriSln 1
Assay Percent Range >99%
Molecular Formula C12H11N3O
Formula Weight 213.24
Synonym 4-Pyridinecarboxylic acid 2-phenylhydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

484710

 
r & d systems
RDS484710
10mg Each for $133.98

484750

 
r & d systems
RDS484750
50mg Each for $567.83
Description & Specifications

Specifications

Formula Weight 213.24
Quantity 10mg
Synonym 4-Pyridinecarboxylic acid 2-phenylhydrazide
Recommended Storage Store at +4°C
Molecular Formula C12H11N3O
CAS 91396-88-2
Assay Percent Range >99%
Chemical Name or Material PluriSln 1
Safety and Handling

Store at +4°C