Invitrogen™ PLK3 Recombinant Human Protein DFS Item
PLK3 is a member of the polo family of serine/threonine kinases.
Manufacturer:  Invitrogen™ LSPV4039
 View more versions of this product
Catalog No. PV4039
Description
Description
PLK3 is involved in the regulation of M phase in the cell cycle.
Specifications
Specifications
NP_004064
Frozen
59.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
PLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PLK3, CNK
Baculovirus
Human
GST Tag
Documents