PLK1 Recombinant Human Protein DFS Item
PLK1 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4038
 View more versions of this product
Catalog No. PV4038
Description
Description
PLK1 is a serine⁄threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis.
Specifications
Specifications
NP_005021
Protein assays and analysis
Frozen
PLK1 Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
PLK1
PLK1 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10 MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 500μg/mL dephosphorylated casein substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
PLK1 Recombinant Human Protein
Human
0.46mg/mL
Frozen
73.3kDa
85% by SDS-PAGE
–80°C
1 Each
PLK1 Recombinant Human Protein
Kinase assay
PLK
62nmoles of phosphate transferred to dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4.2μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.