Please login to your online account to display your discounted pricing

Alfa Aesar™ Plicamycin, 1mg/mL in DMSO, sterile-filtered

Chemical Identifiers

Chemical Name or Material Plicamycin
CAS 18378-89-7
Name Note 1mg/mL in DMSO, sterile-filtered
MDL Number MFCD00135618
Synonym Aureolic acid; Mithramycin A
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67319XF

 
alfa aesar™
J67319XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Plicamycin
MDL Number MFCD00135618
CAS 18378-89-7
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number PZ2800000
Name Note 1mg/mL in DMSO, sterile-filtered
Physical Form Liquid
Synonym Aureolic acid; Mithramycin A
Health Hazard 1 Warning
Storage Note 1 -30° to -10°C
Quantity 1mL
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

-30° to -10°C