missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum wire, For ISA Type R or S Standard Grade Thermocouple, Alfa Aesar™

Form: approx. 4.34g/m Negative leg Type R or S

Manufacturer:  Alfa Aesar 14361BY

 View more versions of this product

Catalog No. AA14361BY



Specifications

Specifications

Platinum
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Wire, 0.508mm (0.02 in.) dia.
White
100cm
Silvery white wire
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
ISA Type R or S Standard

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents