Please login to your online account to display your discounted pricing

Platinum wire, Alfa Aesar™

Formula: 195.08; Form: approx. 1.09g/m

$313.41 - $1,157.19

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Purity Grade For ISA Type R or S Standard Grade
Physical Form Wire, 0.25mm (0.01 in.) dia.
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10287G1

  SDS
alfa aesar
10287G1
1m Each for $313.41

AA10287G5

  SDS
alfa aesar
10287G5
5m Each for $1,157.19
Description & Specifications

Specifications

Quantity 1m
Color Silvery white
Density 21.45g/cm3 at 20°C
EINECS Number 231-116-1
Chemical Name or Material Platinum
Purity Grade For ISA Type R or S Standard Grade
Physical Form Wire, 0.25mm (0.01 in.) dia.
MDL Number MFCD00011179
CAS 7440-06-4
Appearance Silvery white wire
Odor Odorless